Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297548.1 | NP_001284477.1 | 4 | 293 | ||
NM_003432.2 | NP_003423.1 | 4 | 347 | |||
NR_123719.1 | 4 | 495 | ||||
XM_005248359.2 | XP_005248416.1 | 4 | 508 | |||
XM_005248360.2 | XP_005248417.1 | 4 | 305 | |||
XM_005248361.2 | XP_005248418.1 | 4 | 533 | |||
XM_005248362.4 | XP_005248419.1 | 4 | 482 | |||
XM_005248363.4 | XP_005248420.1 | 4 | 478 | |||
XM_005248365.3 | XP_005248422.1 | 4 | 548 | |||
XM_011514123.2 | XP_011512425.1 | 2 | 175 | |||
XM_011514124.2 | XP_011512426.1 | 3 | 190 | |||
XM_017009830.1 | XP_016865319.1 | 4 | 298 | |||
XM_017009831.1 | XP_016865320.1 | 2 | 467 | |||
XM_017009832.1 | XP_016865321.1 | 4 | 533 | |||
XM_017009833.1 | XP_016865322.1 | 4 | 480 | |||
XM_017009834.1 | XP_016865323.1 | 4 | 481 | |||
XM_017009835.1 | XP_016865324.1 | 4 | 323 | |||
XM_017009836.1 | XP_016865325.1 | 4 | 1232 | |||
XM_017009837.1 | XP_016865326.1 | 4 | 560 | |||
XM_017009838.1 | XP_016865327.1 | 2 | 538 | |||
GenBank mRNA | AK057343.1 | - | 4 | 301 | ||
AK299315.1 | - | 4 | 293 | |||
AK307963.1 | - | 4 | 513 | |||
AK307974.1 | - | 48 | ||||
AK310735.1 | - | 4 | 285 | |||
BC035875.2 | - | 1 | 32 | 5519811 | ||
DC326279.1 | - | 4 | 495 | |||
DC371902.1 | - | 4 | 347 |