Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF131 SIRNA Assays ›
Gene Symbol:
ZNF131
Gene Name:
zinc finger protein 131
Gene Aliases:
ZBTB35, pHZ-10
Chromosome Location:
Chr.5: 43120848 - 43176324 on Build GRCh38
UniGene ID:
Hs.535804
Species:
Human
siRNA ID
HSS187808
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF131
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297548.1
NP_001284477.1
5
481
NM_003432.2
NP_003423.1
5
535
NR_123719.1
5
683
NR_123720.1
4
385
XM_005248359.2
XP_005248416.1
5
696
XM_005248360.2
XP_005248417.1
5
493
XM_005248361.2
XP_005248418.1
5
721
XM_005248362.4
XP_005248419.1
5
670
XM_005248363.4
XP_005248420.1
5
666
XM_005248365.3
XP_005248422.1
5
736
XM_005248367.2
XP_005248424.1
4
519
XM_011514123.2
XP_011512425.1
3
363
XM_011514124.2
XP_011512426.1
4
378
XM_017009830.1
XP_016865319.1
5
486
XM_017009831.1
XP_016865320.1
3
655
XM_017009832.1
XP_016865321.1
5
721
XM_017009833.1
XP_016865322.1
5
668
XM_017009834.1
XP_016865323.1
5
669
XM_017009835.1
XP_016865324.1
5
511
XM_017009836.1
XP_016865325.1
5
1420
XM_017009837.1
XP_016865326.1
5
748
XM_017009838.1
XP_016865327.1
3
726
XM_017009839.1
XP_016865328.1
4
602
XM_017009840.1
XP_016865329.1
4
358
XM_017009841.1
XP_016865330.1
4
519
XM_017009842.1
XP_016865331.1
4
525
XM_017009843.1
XP_016865332.1
4
538
GenBank mRNA
AK057343.1
-
5
489
AK299315.1
-
6
481
AK307963.1
-
6
703
AK307974.1
-
237
AK307977.1
-
4
343
BC035875.2
-
2
220
5519811
DC371902.1
-
6
535
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0