Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006763.2 | NP_006754.1 | 1, 2 | 221 | ||
GenBank mRNA | BC105948.1 | - | 1, 2 | 195 | 40032676 | |
BC105949.1 | - | 1, 2 | 195 | 40032675 | ||
U72649.1 | - | 1, 2 | 221 | |||
Y09943.1 | - | 1, 2 | 213 |