Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012293.2 | NP_036425.1 | 7 | 701 | ||
XM_005264707.3 | XP_005264764.1 | 6 | 617 | |||
XM_011510396.1 | XP_011508698.1 | 7 | 458 | |||
XM_011510397.2 | XP_011508699.1 | 6 | 546 | |||
GenBank mRNA | AF200348.1 | - | 7 | 660 | ||
BC030834.1 | - | 7 | 744 | |||
BC098579.1 | - | 7 | 596 | |||
D86983.1 | - | 7 | 660 | |||
EF090903.1 | - | 7 | 660 |