Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198953.1 | NP_001185882.1 | 5 | 564 | ||
NM_024071.3 | NP_076976.1 | 5 | 564 | |||
GenBank mRNA | AK057816.1 | - | 5 | 485 | ||
AK124507.1 | - | 2 | 493 | |||
AK226142.1 | - | 3 | 3527 | |||
AK290519.1 | - | 5 | 477 | |||
AY395756.1 | - | 5 | 479 | |||
BC001130.2 | - | 5 | 485 | 2966683 | ||
BC005999.1 | - | 5 | 500 | 4091809 | ||
BX248261.1 | - | 5 | 470 | |||
CX786145.1 | - | 5 | 564 |