Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201472.1 | NP_001188401.1 | 16 | 1891 | ||
NM_001201473.1 | NP_001188402.1 | 14 | 1690 | |||
NM_024535.4 | NP_078811.3 | 16 | 1945 | |||
GenBank mRNA | AK025674.1 | - | 16 | 1554 | ||
AK094596.1 | - | 16 | 1564 | |||
AK097238.1 | - | 16 | 1411 | |||
AK126176.1 | - | 2 | 717 | |||
AK294045.1 | - | 16 | 1526 | |||
AK296723.1 | - | 16 | 1523 | |||
AK296807.1 | - | 14 | 1356 | |||
AK303353.1 | - | 11 | 1076 | |||
AK309327.1 | - | 16 | 1519 | |||
AK309763.1 | - | 6 | 548 | |||
BC117289.1 | - | 16 | 1564 | 40125840 | ||
BC117291.1 | - | 16 | 1564 | 40125842 |
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 16 | 1945 | ||
GenBank mRNA | AL833954.1 | - | 17 | 1965 |