Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105248.1 | NP_001098718.1 | 19, 20 | 3574 | ||
NM_001105249.1 | NP_001098719.1 | 18, 19 | 3400 | |||
NM_001261841.1 | NP_001248770.1 | 19, 20 | 3529 | |||
NM_001308161.1 | NP_001295090.1 | 18, 19 | 3211 | |||
NM_024780.4 | NP_079056.2 | 15, 16 | 2344 | |||
GenBank mRNA | AK023655.1 | - | 8, 9 | 827 | ||
AK093944.1 | - | 12, 13 | 1401 | |||
AK131202.1 | - | 16, 17 | 2064 | |||
AK299055.1 | - | 18, 19 | 3211 | |||
AK304431.1 | - | 19, 20 | 3529 | |||
AY236494.1 | - | 15, 16 | 2344 | |||
AY263164.1 | - | 15, 16 | 2087 | |||
AY358155.1 | - | 16, 17 | 2749 | |||
BC027602.1 | - | 15, 16 | 2299 | 4839111 | ||
BC038118.2 | - | 15, 16 | 2039 | 5265527 | ||
CR749359.1 | - | 17, 18 | 2229 |