Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024789.3 | NP_079065.2 | 8 | 1295 | ||
XM_006717973.3 | XP_006718036.1 | 10 | 1498 | |||
XM_006717974.3 | XP_006718037.1 | 7 | 1234 | |||
XM_011540160.2 | XP_011538462.1 | 9 | 1918 | |||
XM_011540163.2 | XP_011538465.1 | 9 | 1408 | |||
XM_011540167.2 | XP_011538469.1 | 6 | 721 | |||
XM_011540169.2 | XP_011538471.1 | 7 | 909 | |||
XM_017016654.1 | XP_016872143.1 | 9 | 1452 | |||
XM_017016655.1 | XP_016872144.1 | 8 | 1496 | |||
XM_017016656.1 | XP_016872145.1 | 9 | 1558 | |||
XM_017016657.1 | XP_016872146.1 | 9 | 1918 | |||
XM_017016658.1 | XP_016872147.1 | 8 | 1362 | |||
XM_017016659.1 | XP_016872148.1 | 8 | 1465 | |||
GenBank mRNA | AK026182.1 | - | 4 | 501 | ||
AK296583.1 | - | 8 | 1156 | |||
AK301612.1 | - | 7 | 852 | |||
BC057787.1 | - | 4 | 669 | |||
BC067530.1 | - | 3 | 283 | |||
BC067531.1 | - | 3 | 283 | |||
BC113589.1 | - | 6 | 1110 | 8322641 |