Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
siRNA ID
HSS188245
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
16
2368
NM_001160104.1
NP_001153576.1
16
2353
NM_001326295.1
NP_001313224.1
14
1978
NM_001326296.1
NP_001313225.1
15
2293
NM_001326297.1
NP_001313226.1
16
2552
NM_001326298.1
NP_001313227.1
13
1903
NM_001326299.1
NP_001313228.1
15
2278
NM_001326300.1
NP_001313229.1
15
2256
NM_001326301.1
NP_001313230.1
15
2477
NM_001326302.1
NP_001313231.1
15
2253
NM_001326303.1
NP_001313232.1
13
2081
NM_001326304.1
NP_001313233.1
13
1860
NM_001326305.1
NP_001313234.1
14
2181
NM_001326306.1
NP_001313235.1
12
1744
NM_001326307.1
NP_001313236.1
15
2296
NM_001326308.1
NP_001313237.1
13
1863
NM_001326309.1
NP_001313238.1
12
1788
NM_001326310.1
NP_001313239.1
16
2356
NM_001326311.1
NP_001313240.1
12
1785
NM_001326312.1
NP_001313241.1
15
2281
NM_001326313.1
NP_001313242.1
12
1747
NM_001326314.1
NP_001313243.1
14
2178
NM_001326315.1
NP_001313244.1
16
2549
NM_001326316.1
NP_001313245.1
16
2216
NM_024824.4
NP_079100.2
16
2371
NM_207660.3
NP_997543.1
13
1900
NM_207661.2
NP_997544.1
14
1826
NM_207662.3
NP_997545.2
6
1197
NR_136936.1
15
2211
XM_005268067.4
XP_005268124.1
16
2351
XM_005268068.4
XP_005268125.1
15
2291
XM_005268069.4
XP_005268126.1
14
1973
XM_005268070.4
XP_005268127.1
14
1970
XM_005268071.4
XP_005268128.1
13
1883
XM_005268073.3
XP_005268130.1
8
1593
XM_006720257.2
XP_006720320.1
7
1518
XM_011537160.2
XP_011535462.1
16
2479
XM_011537161.2
XP_011535463.1
15
2210
XM_017021648.1
XP_016877137.1
15
2288
XM_017021649.1
XP_016877138.1
15
2276
XM_017021650.1
XP_016877139.1
15
2273
GenBank mRNA
AF155107.1
-
5
459
AF474376.1
-
5
1066
AK021868.1
-
13
1747
AK090809.1
-
2
1431
AK302136.1
-
13
1718
AK303123.1
-
15
1819
AL834215.1
-
14
1826
AY578060.1
-
16
2255
AY578061.1
-
13
1839
AY578062.1
-
14
1826
AY578063.1
-
8
904
BC011793.1
-
16
2237
4298961
BC023641.2
-
12
1874
BC027607.2
-
6
1183
BX248265.1
-
12
1890
BX248281.1
-
8
972
BX248769.1
-
3
261
BX640716.1
-
16
2250
BX640876.1
-
5
564
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0