Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP8B4 SIRNA Assays ›
Gene Symbol:
ATP8B4
Gene Name:
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases:
ATPIM
Chromosome Location:
Chr.15: 49858238 - 50182096 on Build GRCh38
UniGene ID:
Hs.511311
Species:
Human
siRNA ID
HSS188260
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP8B4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024837.3
NP_079113.2
23
2667
NR_073596.1
23
2566
NR_073597.1
22
2519
NR_073598.1
24
2774
XM_011522046.2
XP_011520348.1
25
3056
XM_011522047.2
XP_011520349.1
24
3581
XM_011522048.1
XP_011520350.1
24
2854
XM_011522049.2
XP_011520351.1
24
2964
XM_011522051.2
XP_011520353.1
25
2891
XM_011522052.2
XP_011520354.1
24
2949
XM_011522053.1
XP_011520355.1
24
2730
XM_011522056.2
XP_011520358.2
24
3116
XM_011522058.2
XP_011520360.1
20
3325
XM_011522059.1
XP_011520361.1
19
2346
XM_011522060.1
XP_011520362.1
22
2581
XM_011522061.1
XP_011520363.1
21
2496
XM_011522062.1
XP_011520364.1
19
2335
XM_011522063.1
XP_011520365.1
19
2317
XM_011522064.1
XP_011520366.1
15
2018
XM_011522069.2
XP_011520371.1
14
1839
XM_011522070.1
XP_011520372.1
10
1259
XM_017022587.1
XP_016878076.1
23
3032
XM_017022588.1
XP_016878077.1
23
2957
XM_017022589.1
XP_016878078.1
23
2927
XM_017022590.1
XP_016878079.1
25
2868
XM_017022591.1
XP_016878080.1
24
2815
XM_017022592.1
XP_016878081.1
23
2878
XM_017022593.1
XP_016878082.1
24
3116
XM_017022594.1
XP_016878083.1
20
2376
XM_017022595.1
XP_016878084.1
20
4381
XM_017022596.1
XP_016878085.1
15
2009
GenBank mRNA
AB075819.1
-
24
2774
AK123747.1
-
4
433
AK128602.1
-
4
881
BC144297.1
-
22
2515
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
phospholipid translocation
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0