Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
siRNA ID
HSS188299
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
3
358
NM_001272105.1
NP_001259034.1
4
430
NM_001272106.1
NP_001259035.1
2
240
NM_001272107.1
NP_001259036.1
3
358
NM_001272108.1
NP_001259037.1
3
316
NM_001272109.1
NP_001259038.1
2
240
NM_001272110.1
NP_001259039.1
3
358
NM_001272111.1
NP_001259040.1
3
316
NM_001272112.1
NP_001259041.1
2
240
NM_001272113.1
NP_001259042.1
3
316
NM_024897.3
NP_079173.2
3
358
NM_198406.2
NP_940798.1
3
316
NR_073610.1
2
240
XM_005245494.3
XP_005245551.1
2
611
XM_006711546.2
XP_006711609.1
3
256
XM_006711547.3
XP_006711610.1
3
729
XM_006711548.3
XP_006711611.1
2
611
XM_006711552.2
XP_006711615.1
3
687
XM_006711553.3
XP_006711616.1
3
729
XM_011510000.2
XP_011508302.1
3
687
XM_017002385.1
XP_016857874.1
2
1887
GenBank mRNA
AF455045.1
-
3
277
AF455046.1
-
2
184
AF455047.1
-
2
154
AK054682.1
-
3
297
AK094346.1
-
3
255
AK095143.1
-
3
254
AK095197.1
-
3
293
AK294143.1
-
2
168
AK295915.1
-
2
182
AK295916.1
-
2
196
AK307441.1
-
1
20
AK316034.1
-
4
369
AL834227.1
-
3
249
AY424284.1
-
3
237
BC058509.1
-
3
200
5770270
BI551776.1
-
3
260
BP208594.1
-
3
254
BP217276.1
-
2
240
BP362806.1
-
3
248
DT218672.1
-
4
371
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0