Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024939.2 | NP_079215.2 | 7 | 976 | ||
XM_005256153.4 | XP_005256210.1 | 7 | 915 | |||
XM_006721273.3 | XP_006721336.1 | 7 | 984 | |||
XM_011523338.2 | XP_011521640.1 | 6 | 876 | |||
GenBank mRNA | AF289577.1 | - | 1 | 79 | ||
AK025571.1 | - | 7 | 905 | |||
BC030146.2 | - | 7 | 798 | 4810948 | ||
BC069241.1 | - | 7 | 1327 | |||
LN680438.1 | - | 7 | 788 | |||
LN680439.1 | - | 7 | 788 |