Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287181.1 | NP_001274110.1 | 3, 4 | 366 | ||
NM_025055.4 | NP_079331.3 | 13, 14 | 1969 | |||
NM_182791.3 | NP_877592.2 | 3, 4 | 366 | |||
XM_005254692.1 | XP_005254749.1 | 13, 14 | 1969 | |||
XM_011522085.2 | XP_011520387.1 | 14, 15 | 2814 | |||
XM_011522086.1 | XP_011520388.1 | 13, 14 | 1662 | |||
XM_011522087.2 | XP_011520389.1 | 14, 15 | 1800 | |||
XM_011522088.1 | XP_011520390.1 | 13, 14 | 1670 | |||
XM_011522089.2 | XP_011520391.1 | 7, 8 | 960 | |||
XM_011522090.2 | XP_011520392.2 | 3, 4 | 664 | |||
XM_017022623.1 | XP_016878112.1 | 14, 15 | 2815 | |||
XM_017022624.1 | XP_016878113.1 | 14, 15 | 2814 | |||
XM_017022625.1 | XP_016878114.1 | 14, 15 | 2814 | |||
XM_017022626.1 | XP_016878115.1 | 14, 15 | 2814 | |||
XM_017022627.1 | XP_016878116.1 | 14, 15 | 2814 | |||
XM_017022628.1 | XP_016878117.1 | 14, 15 | 2814 | |||
XM_017022629.1 | XP_016878118.1 | 16, 17 | 2555 | |||
XM_017022632.1 | XP_016878121.1 | 3, 4 | 499 | |||
GenBank mRNA | AK026821.1 | - | 3, 4 | 348 | ||
AK057417.1 | - | 13, 14 | 1855 | |||
AK290709.1 | - | 3, 4 | 366 | |||
AK310852.1 | - | 3, 4 | 366 | |||
BC031684.1 | - | 10, 11 | 1179 | |||
BP292996.1 | - | 3, 4 | 348 |