Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324282.1 | NP_001311211.1 | 8 | 971 | ||
NM_001324283.1 | NP_001311212.1 | 8 | 982 | |||
NM_001324284.1 | NP_001311213.1 | 10 | 1171 | |||
NM_001324285.1 | NP_001311214.1 | 10 | 1179 | |||
NM_001324286.1 | NP_001311215.1 | 6 | 810 | |||
NM_001324287.1 | NP_001311216.1 | 7 | 895 | |||
NM_001324288.1 | NP_001311217.1 | 10 | 1210 | |||
NM_001324289.1 | NP_001311218.1 | 9 | 1125 | |||
NM_003473.3 | NP_003464.1 | 9 | 1067 | |||
XM_011519695.2 | XP_011517997.1 | 9 | 1043 | |||
XM_017016685.1 | XP_016872174.1 | 5 | 1112 | |||
GenBank mRNA | AK303083.1 | - | 7 | 755 | ||
AK314405.1 | - | 9 | 951 | |||
BC030586.2 | - | 9 | 1010 | 4820857 | ||
BX647982.1 | - | 3 | 152 | |||
U43899.1 | - | 9 | 894 |