Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271539.1 | NP_001258468.1 | 16 | 1549 | ||
NR_073206.1 | 18 | 1836 | ||||
NR_073207.1 | 16 | 1717 | ||||
XM_006723646.2 | XP_006723709.1 | 17 | 1717 | |||
XM_006723647.2 | XP_006723710.1 | 17 | 1717 | |||
XM_006723648.2 | XP_006723711.1 | 16 | 1660 | |||
XM_006723650.2 | XP_006723713.1 | 17 | 1717 | |||
XM_006723651.2 | XP_006723714.1 | 17 | 1717 | |||
XM_006723652.2 | XP_006723715.1 | 16 | 1551 | |||
XM_006723654.3 | XP_006723717.1 | 8 | 732 | |||
XM_011529374.1 | XP_011527676.1 | 16 | 1657 | |||
XM_011529375.1 | XP_011527677.1 | 17 | 1606 | |||
XM_011529376.1 | XP_011527678.1 | 16 | 1549 | |||
XM_011529377.1 | XP_011527679.1 | 18 | 1926 | |||
XM_011529378.1 | XP_011527680.1 | 15 | 1435 | |||
XM_011529379.1 | XP_011527681.1 | 15 | 1468 | |||
XM_017028084.1 | XP_016883573.1 | 6 | 654 | |||
GenBank mRNA | AK302170.1 | - | 16 | 1546 | ||
AL137678.1 | - | 16 | 1601 | |||
DA857535.1 | - | 3 | 192 |