Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171091.1 | NP_001164562.1 | 3, 4 | 302 | ||
NM_025241.2 | NP_079517.1 | 3, 4 | 404 | |||
XM_017027325.1 | XP_016882814.1 | 4, 5 | 835 | |||
GenBank mRNA | AF272893.1 | - | 3, 4 | 403 | ||
AF272894.1 | - | 3, 4 | 293 | |||
BC008288.1 | - | 3, 4 | 302 | 3659797 | ||
BM764980.1 | - | 3, 4 | 404 |