Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 4 | 1041 | ||
NM_182683.2 | NP_872624.1 | 5 | 876 | |||
NM_182684.2 | NP_872625.1 | 6 | 956 | |||
GenBank mRNA | AK056760.1 | - | 5 | 684 | ||
AK290096.1 | - | 4 | 944 | |||
AY233462.1 | - | 6 | 789 | |||
BC004304.1 | - | 4 | 865 | 3625247 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 4 | 1362 | ||
XM_006716140.3 | XP_006716203.1 | 6 | 1034 | |||
XM_006716141.3 | XP_006716204.1 | 5 | 954 |