Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005089.3 | NP_005080.1 | 10 | 924 | ||
XM_005274597.2 | XP_005274654.2 | 10 | 892 | |||
XM_011545589.2 | XP_011543891.2 | 9 | 856 | |||
XM_011545590.2 | XP_011543892.2 | 10 | 1694 | |||
XM_017029881.1 | XP_016885370.1 | 10 | 891 | |||
XM_017029882.1 | XP_016885371.1 | 9 | 846 | |||
XM_017029883.1 | XP_016885372.1 | 10 | 987 | |||
GenBank mRNA | AK309673.1 | - | 10 | 891 | ||
BC050451.1 | - | 10 | 895 | 6499970 | ||
BC113454.1 | - | 10 | 924 | 8322506 | ||
BC113480.1 | - | 10 | 924 | 8322532 | ||
D49677.1 | - | 10 | 903 |
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK299902.1 | - | 4 | 672 | ||
AK310729.1 | - | 3 | 772 |
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204199.1 | NP_001191128.1 | 5 | 1421 |
Gene Symbol: ZRSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104811.1 | - | 1 | 922 | ||
BC113599.1 | - | 1 | 920 | |||
D49676.1 | - | 1 | 918 |