Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP9X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039590.2 | NP_001034679.2 | 40 | 7533 | ||
NM_001039591.2 | NP_001034680.2 | 40 | 7533 | |||
XM_005272675.4 | XP_005272732.1 | 40 | 7763 | |||
XM_005272676.4 | XP_005272733.1 | 40 | 7717 | |||
GenBank mRNA | AB209666.1 | - | 11 | 2521 | ||
AF070645.1 | - | 2 | 225 | |||
BC046205.1 | - | 4 | 659 | |||
BC063645.1 | - | 9 | 1994 | |||
X98296.1 | - | 39 | 6938 |