Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195388.1 | NP_001182317.1 | 7 | 1489 | ||
NM_031281.2 | NP_112571.2 | 7 | 1489 | |||
XM_011510030.1 | XP_011508332.1 | 6 | 1191 | |||
XM_011510031.1 | XP_011508333.1 | 7 | 1487 | |||
XM_011510032.1 | XP_011508334.1 | 6 | 1208 | |||
XM_011510033.1 | XP_011508335.1 | 7 | 1487 | |||
GenBank mRNA | AF343662.1 | - | 7 | 1424 | ||
AF343663.1 | - | 7 | 1424 | |||
AF343664.1 | - | 7 | 1424 | |||
AF369794.2 | - | 7 | 1489 | |||
AF397453.1 | - | 7 | 1331 | |||
AK300010.1 | - | 6 | 1215 | |||
AK301356.1 | - | 8 | 1529 | |||
BC101066.2 | - | 7 | 1359 | 40008649 | ||
BC101067.1 | - | 7 | 1359 | 40008653 | ||
BC101069.2 | - | 7 | 1359 | 40008658 |