Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031284.4 | NP_112574.3 | 7 | 1222 | ||
NR_023318.1 | 7 | 1435 | ||||
NR_023319.1 | 5 | 936 | ||||
XM_006720712.3 | XP_006720775.1 | 7 | 1225 | |||
XM_006720713.3 | XP_006720776.1 | 7 | 1386 | |||
XM_011522102.2 | XP_011520404.1 | 4 | 808 | |||
XM_011522103.2 | XP_011520405.1 | 4 | 805 | |||
XM_017022664.1 | XP_016878153.1 | 3 | 1166 | |||
GenBank mRNA | AF204270.1 | - | 6 | 814 | ||
AK055526.1 | - | 5 | 927 | |||
AK057651.1 | - | 1 | 1154 | |||
AK075560.1 | - | 7 | 1157 | |||
AK293282.1 | - | 7 | 917 | |||
AK294391.1 | - | 6 | 962 | |||
AL136873.1 | - | 4 | 485 | |||
AL832254.1 | - | 2 | 3243 | |||
BC006112.1 | - | 7 | 1167 | 3347312 | ||
BC044589.1 | - | 7 | 1381 | |||
CR533447.1 | - | 3 | 306 |