Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258426.1 | NP_001245355.1 | 9 | 1171 | ||
NM_001258427.1 | NP_001245356.1 | 9 | 1204 | |||
NM_001258428.1 | NP_001245357.1 | 9 | 1276 | |||
NM_031467.2 | NP_113655.2 | 9 | 1204 | |||
XM_005268521.3 | XP_005268578.2 | 9 | 1761 | |||
XM_011537694.2 | XP_011535996.1 | 9 | 1833 | |||
XM_017009933.1 | XP_016865422.1 | 9 | 1833 | |||
XM_017009934.1 | XP_016865423.1 | 9 | 1833 | |||
XM_017009935.1 | XP_016865424.1 | 9 | 1800 | |||
XM_017009936.1 | XP_016865425.1 | 9 | 1761 | |||
XM_017009937.1 | XP_016865426.1 | 8 | 1719 | |||
XM_017009938.1 | XP_016865427.1 | 9 | 1833 | |||
XM_017009939.1 | XP_016865428.1 | 9 | 1728 | |||
XM_017009940.1 | XP_016865429.1 | 9 | 1833 | |||
XM_017009941.1 | XP_016865430.1 | 9 | 1833 | |||
GenBank mRNA | AB032762.1 | - | 9 | 1163 | ||
AF313465.1 | - | 9 | 1263 | |||
AF332961.1 | - | 9 | 1204 | |||
AF336237.1 | - | 9 | 1163 | |||
BC136262.1 | - | 9 | 1195 | 9020249 | ||
BC143602.1 | - | 9 | 1195 | 9052115 |