Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320823.1 | NP_001307752.1 | 9 | 897 | ||
NM_001320824.1 | NP_001307753.1 | 9 | 952 | |||
NM_001320825.1 | NP_001307754.1 | 8 | 696 | |||
NM_032118.3 | NP_115494.1 | 9 | 919 | |||
XM_017005064.1 | XP_016860553.1 | 8 | 698 | |||
GenBank mRNA | AI203710.1 | - | 2 | 247 | ||
AK023015.1 | - | 9 | 912 | |||
AK222696.1 | - | 9 | 912 | |||
AK311621.1 | - | 4 | 498 | |||
BC051848.1 | - | 9 | 884 | 6150670 | ||
CD249537.1 | - | 8 | 712 |