Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 7 | 979 | ||
NM_199179.3 | NP_954648.3 | 6 | 829 | |||
NM_199180.3 | NP_954649.3 | 7 | 1121 | |||
XM_011527362.1 | XP_011525664.1 | 8 | 1181 | |||
XM_011527363.1 | XP_011525665.1 | 8 | 1162 | |||
XM_011527365.1 | XP_011525667.1 | 7 | 1031 | |||
XM_011527366.1 | XP_011525668.1 | 6 | 1292 | |||
XM_017027348.1 | XP_016882837.1 | 7 | 1688 | |||
GenBank mRNA | AB593116.1 | - | 7 | 1085 | ||
AK307865.1 | - | 7 | 1442 | |||
AL136654.1 | - | 7 | 1105 | |||
AY305301.1 | - | 7 | 923 | |||
AY305302.1 | - | 6 | 773 | |||
AY358742.1 | - | 7 | 1107 | |||
BC064925.1 | - | 10 | 1044 | 6141879 |