Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103511.1 | 7, 8 | 1058 | |||
XM_017007311.1 | XP_016862800.1 | 8, 9 | 2641 | |||
GenBank mRNA | AK292958.1 | - | 6, 7 | 722 | ||
AL136815.1 | - | 5, 6 | 489 | |||
BC024239.2 | - | 7, 8 | 828 | 4826873 | ||
BU607306.1 | - | 2, 3 | 577 |