Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032164.2 | NP_115540.2 | 1 | 362 | ||
XM_017012710.1 | XP_016868199.1 | 1 | 410 | |||
XM_017012711.1 | XP_016868200.1 | 1 | 410 | |||
GenBank mRNA | AK022360.1 | - | 1 | 405 | ||
AK172865.1 | - | 1 | 375 | |||
AK222948.1 | - | 1 | 355 | |||
AY261374.1 | - | 1 | 405 | |||
AY642122.1 | - | 1 | 389 | |||
BC017051.1 | - | 1 | 372 | |||
BC025241.1 | - | 1 | 389 | 4641268 | ||
BC051847.1 | - | 1 | 362 | 6007172 |