Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP48 SIRNA Assays ›
Gene Symbol:
USP48
Gene Name:
ubiquitin specific peptidase 48
Gene Aliases:
RAP1GA1, USP31
Chromosome Location:
Chr.1: 21678298 - 21783620 on Build GRCh38
UniGene ID:
Hs.467524
Species:
Human
siRNA ID
HSS188955
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP48
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032730.1
NP_001027902.1
10, 11
1529
NM_032236.5
NP_115612.4
10, 11
1529
XM_005246009.2
XP_005246066.1
10, 11
1533
XM_006710951.2
XP_006711014.2
10, 11
1530
XM_006710955.3
XP_006711018.1
10, 11
1509
XM_011542262.2
XP_011540564.1
10, 11
1423
XM_011542263.2
XP_011540565.1
10, 11
1426
XM_011542264.1
XP_011540566.1
9, 10
1123
XM_011542265.2
XP_011540567.1
11, 12
2089
XM_011542267.2
XP_011540569.1
10, 11
1442
XM_011542268.2
XP_011540570.1
2, 3
142
XM_017002495.1
XP_016857984.1
10, 11
1530
XM_017002496.1
XP_016857985.1
10, 11
1533
XM_017002497.1
XP_016857986.1
2, 3
187
GenBank mRNA
AF502942.1
-
10, 11
1431
AK074247.1
-
10, 11
1423
AK226150.1
-
10, 11
1525
AK315005.1
-
10, 11
1309
BC007326.2
-
1, 2
97
BC067261.1
-
10, 11
1497
BC103724.1
-
10, 11
1471
BC104896.1
-
10, 11
1325
8143899
BC143343.1
-
10, 11
1325
9051848
BC143349.1
-
10, 11
1325
9051854
BC143354.1
-
10, 11
1322
9051859
BM462540.1
-
1, 2
103
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
protein deubiquitination
thiol-dependent ubiquitin-specific protease activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0