Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 3 | 654 | ||
NM_020912.1 | NP_065963.1 | 3 | 654 | |||
NM_032296.2 | NP_115672.2 | 3 | 654 | |||
XM_006720962.3 | XP_006721025.1 | 3 | 756 | |||
XM_011522701.2 | XP_011521003.1 | 3 | 758 | |||
XM_011522702.2 | XP_011521004.1 | 3 | 1232 | |||
XM_011522703.2 | XP_011521005.1 | 4 | 853 | |||
XM_011522704.2 | XP_011521006.1 | 3 | 759 | |||
XM_011522705.2 | XP_011521007.1 | 3 | 757 | |||
XM_011522707.2 | XP_011521009.1 | 3 | 760 | |||
XM_017023772.1 | XP_016879261.1 | 5 | 950 | |||
GenBank mRNA | AB046772.1 | - | 3 | 634 | ||
BC028572.1 | - | 3 | 655 | 4839062 |