Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270041.1 | NP_001256970.1 | 4 | 531 | ||
NM_001270042.1 | NP_001256971.1 | 4 | 544 | |||
NM_032372.5 | NP_115748.1 | 5 | 684 | |||
NR_070308.1 | 6 | 1062 | ||||
NR_070309.1 | 5 | 844 | ||||
XM_011540267.2 | XP_011538569.1 | 4 | 503 | |||
XM_011540268.1 | XP_011538570.1 | 4 | 493 | |||
GenBank mRNA | AK093378.1 | - | 5 | 652 | ||
BC007374.2 | - | 5 | 830 | 3637472 | ||
BC018606.2 | - | 6 | 1062 | |||
BQ229460.1 | - | 4 | 555 |