Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278636.1 | NP_001265565.1 | 5 | 531 | ||
NM_032565.4 | NP_115954.1 | 4 | 469 | |||
NR_103802.1 | 5 | 590 | ||||
NR_103803.1 | 3 | 399 | ||||
GenBank mRNA | AF243433.1 | - | 4 | 458 | ||
BC018478.1 | - | 4 | 435 | 4476654 | ||
BC021021.1 | - | 5 | 508 | |||
BC073152.1 | - | 5 | 590 | |||
BC092471.1 | - | 4 | 464 | 4796715 | ||
BY799168.2 | - | 3 | 365 | |||
CA437822.1 | - | 1 | 523 |