Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321209.1 | NP_001308138.1 | 4 | 350 | ||
NM_001321210.1 | NP_001308139.1 | 4 | 350 | |||
NM_032574.3 | NP_115963.1 | 4 | 490 | |||
NR_135585.1 | 4 | 343 | ||||
XM_006712117.3 | XP_006712180.1 | 4 | 355 | |||
XM_006712118.3 | XP_006712181.1 | 4 | 355 | |||
XM_011533133.2 | XP_011531435.1 | 4 | 423 | |||
XM_017005111.1 | XP_016860600.1 | 4 | 355 | |||
GenBank mRNA | AF226998.1 | - | 4 | 409 | ||
AV725116.1 | - | 4 | 281 | |||
BC015970.1 | - | 4 | 312 | 4081996 | ||
BG940527.1 | - | 4 | 298 | |||
BI752326.1 | - | 4 | 311 | |||
BM505380.1 | - | 2 | 393 | |||
BQ276899.1 | - | 350 |