Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM25 SIRNA Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.11: 118531088 - 118546670 on Build GRCh38
UniGene ID:
Hs.564188
Species:
Human
siRNA ID
HSS189230
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144034.1
NP_001137506.1
4
575
NM_001144035.1
NP_001137507.1
4
572
NM_001144036.1
NP_001137508.1
3
263
NM_001144037.1
NP_001137509.1
4
575
NM_001144038.1
NP_001137510.1
4
575
NM_001318755.1
NP_001305684.1
4
575
NM_032780.3
NP_116169.2
4
575
NR_134852.1
2
166
XM_005271703.1
XP_005271760.1
4
484
XM_006718926.2
XP_006718989.1
4
459
XM_006718927.2
XP_006718990.1
4
459
XM_011543037.2
XP_011541339.1
4
459
XM_011543038.2
XP_011541340.1
4
459
XM_017018422.1
XP_016873911.1
4
484
XM_017018423.1
XP_016873912.1
4
484
XM_017018424.1
XP_016873913.1
4
365
XM_017018425.1
XP_016873914.1
4
388
XM_017018426.1
XP_016873915.1
4
484
XM_017018427.1
XP_016873916.1
4
365
XM_017018428.1
XP_016873917.1
4
388
XM_017018429.1
XP_016873918.1
4
365
XM_017018430.1
XP_016873919.1
4
459
XM_017018431.1
XP_016873920.1
4
459
XM_017018432.1
XP_016873921.1
4
365
XM_017018433.1
XP_016873922.1
4
388
GenBank mRNA
AK027305.1
-
4
464
AK056325.1
-
3
149
AK075437.1
-
4
474
AK096260.1
-
2
1279
AK124814.1
-
3
776
AK225415.1
-
4
461
AK292359.1
-
4
459
AK297255.1
-
4
461
AY358919.1
-
4
460
BC042896.1
-
4
454
5168151
BC051841.1
-
3
443
5725777
BF513120.1
-
4
575
DA760733.1
-
2
52
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0