Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079669.2 | NP_001073137.1 | 2 | 280 | ||
NM_001286453.1 | NP_001273382.1 | 2 | 280 | |||
NM_032813.3 | NP_116202.2 | 3 | 490 | |||
XM_011521121.2 | XP_011519423.1 | 3 | 468 | |||
XM_011521122.2 | XP_011519424.1 | 3 | 384 | |||
XM_011521123.2 | XP_011519425.1 | 3 | 545 | |||
XM_011521124.2 | XP_011519426.1 | 2 | 173 | |||
XM_011521125.1 | XP_011519427.1 | 3 | 377 | |||
XM_017020796.1 | XP_016876285.1 | 3 | 545 | |||
XM_017020797.1 | XP_016876286.1 | 2 | 198 | |||
XM_017020798.1 | XP_016876287.1 | 3 | 468 | |||
XM_017020799.1 | XP_016876288.1 | 2 | 153 | |||
XM_017020800.1 | XP_016876289.1 | 3 | 377 | |||
GenBank mRNA | AK056409.1 | - | 3 | 350 | ||
AK290345.1 | - | 2 | 140 | |||
AK299859.1 | - | 2 | 119 | |||
BX647956.1 | - | 2 | 171 | |||
BX648008.1 | - | 2 | 124 |