Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032873.4 | NP_116262.2 | 9 | 1621 | ||
XM_005271712.3 | XP_005271769.1 | 9 | 3867 | |||
XM_011543041.2 | XP_011541343.1 | 9 | 1259 | |||
XM_011543042.1 | XP_011541344.1 | 9 | 1625 | |||
GenBank mRNA | AB075839.1 | - | 9 | 1473 | ||
AF425252.1 | - | 7 | 918 | |||
AK075203.1 | - | 9 | 1589 | |||
AK222843.1 | - | 9 | 1571 | |||
AK222846.1 | - | 9 | 1571 | |||
BC007541.1 | - | 9 | 1575 | 2958242 |