Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC43A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198810.1 | NP_001185739.1 | 8 | 855 | ||
NM_003627.5 | NP_003618.1 | 8 | 1026 | |||
XM_005274358.4 | XP_005274415.1 | 9 | 1166 | |||
XM_011545320.2 | XP_011543622.1 | 8 | 1075 | |||
XM_011545321.2 | XP_011543623.1 | 6 | 642 | |||
XM_011545322.1 | XP_011543624.1 | 5 | 491 | |||
XM_017018451.1 | XP_016873940.1 | 9 | 1054 | |||
XM_017018452.1 | XP_016873941.1 | 7 | 995 | |||
XM_017018453.1 | XP_016873942.1 | 6 | 832 | |||
GenBank mRNA | AB103033.1 | - | 8 | 993 | ||
AF045584.1 | - | 8 | 813 | |||
AK027732.1 | - | 6 | 591 | |||
BC001639.2 | - | 8 | 855 | 2959372 |