Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303486.1 | NP_001290415.1 | 2, 3 | 146 | ||
NM_001324381.1 | NP_001311310.1 | 2, 3 | 150 | |||
NM_003631.3 | NP_003622.2 | 2, 3 | 576 | |||
NR_130168.1 | 2, 3 | 576 | ||||
NR_136755.1 | 2, 3 | 576 | ||||
NR_136756.1 | 2, 3 | 576 | ||||
XM_011540305.1 | XP_011538607.1 | 2, 3 | 542 | |||
XM_011540308.2 | XP_011538610.1 | 2, 3 | 542 | |||
GenBank mRNA | AF005043.1 | - | 3, 4 | 440 | ||
AK302560.1 | - | 2, 3 | 117 | |||
AK307788.1 | - | 2, 3 | 527 | |||
AY258587.1 | - | 2, 3 | 535 | |||
AY575848.1 | - | 2, 3 | 130 | |||
BC050560.1 | - | 2, 3 | 502 | 6064831 | ||
BC052966.1 | - | 2, 3 | 490 | 6042331 | ||
DQ867088.1 | - | 2, 3 | 354 |