Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145426.1 | NP_001138898.1 | 2 | 550 | ||
NM_003651.4 | NP_003642.3 | 2 | 550 | |||
XM_011520871.2 | XP_011519173.1 | 2 | 644 | |||
XM_017020122.1 | XP_016875611.1 | 2 | 644 | |||
XM_017020123.1 | XP_016875612.1 | 2 | 644 | |||
GenBank mRNA | AB209896.1 | - | 2 | 107 | ||
AK314846.1 | - | 2 | 519 | |||
AU117252.1 | - | 2 | 550 | |||
BC008801.2 | - | 2 | 508 | 3634981 | ||
BC009744.2 | - | 2 | 503 | 3835480 | ||
BC015564.1 | - | 2 | 499 | 4137643 | ||
BC015913.1 | - | 2 | 492 | 4563632 | ||
BC021926.1 | - | 2 | 501 | 4551107 | ||
M24069.1 | - | 2 | 517 | |||
X72712.1 | - | 2 | 528 | |||
X95325.1 | - | 2 | 490 |
Gene Symbol: YBX3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027011.1 | 1 | 331 | |||
GenBank mRNA | BI826429.1 | - | 1 | 39 | ||
DB451883.1 | - | 1 | 302 |