Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: API5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142930.1 | NP_001136402.1 | 13 | 1545 | ||
NM_001142931.1 | NP_001136403.1 | 12 | 1383 | |||
NM_001243747.1 | NP_001230676.1 | 10 | 1026 | |||
NM_006595.3 | NP_006586.1 | 13 | 1545 | |||
NR_024625.1 | 14 | 1607 | ||||
XM_006718359.3 | XP_006718422.1 | 13 | 1535 | |||
XM_017018464.1 | XP_016873953.1 | 14 | 1592 | |||
XM_017018465.1 | XP_016873954.1 | 14 | 1592 | |||
GenBank mRNA | AF229253.1 | - | 12 | 1159 | ||
AF229254.1 | - | 13 | 1485 | |||
AK293303.1 | - | 10 | 927 | |||
AK294724.1 | - | 12 | 1340 | |||
AK299288.1 | - | 13 | 1511 | |||
AK304157.1 | - | 14 | 1573 | |||
AY311389.1 | - | 13 | 1484 | |||
BC017709.1 | - | 13 | 1465 | 4419401 | ||
BT007093.1 | - | 13 | 1372 | |||
U83857.1 | - | 13 | 1467 | |||
Y15906.1 | - | 13 | 1504 |