Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033426.2 | NP_219494.2 | 3 | 467 | ||
GenBank mRNA | AB051524.1 | - | 2 | 342 | ||
AK096603.1 | - | 3 | 384 | |||
AK315124.1 | - | 3 | 444 | |||
AL834514.1 | - | 3 | 370 | |||
BC037300.2 | - | 3 | 387 | 4827858 | ||
BI546511.1 | - | 3 | 504 |