Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286957.1 | NP_001273886.1 | 1 | 275 | ||
NM_033124.4 | NP_149115.2 | 1 | 275 | |||
GenBank mRNA | AF382188.1 | - | 1 | 240 | ||
AK093051.1 | - | 1 | 248 | |||
AK302082.1 | - | 1 | 252 | |||
AK314623.1 | - | 1 | 220 | |||
BC039317.1 | - | 1 | 214 | 5271866 | ||
DB026303.1 | - | 1 | 275 |