Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK10 SIRNA Assays ›
Gene Symbol:
CDK10
Gene Name:
cyclin dependent kinase 10
Gene Aliases:
PISSLRE
Chromosome Location:
Chr.16: 89686612 - 89696364 on Build GRCh38
UniGene ID:
Hs.699177
Species:
Human
siRNA ID
HSS189424
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098533.2
NP_001092003.2
3
197
NM_001160367.1
NP_001153839.1
3
197
NM_052987.3
NP_443713.2
3
197
NM_052988.4
NP_443714.3
3
235
NR_027702.1
2
162
NR_027703.1
2
162
XM_006721308.2
XP_006721371.1
3
291
XM_006721310.2
XP_006721373.1
3
291
XM_011523405.2
XP_011521707.1
3
291
XM_011523406.2
XP_011521708.1
3
291
XM_011523407.2
XP_011521709.1
3
291
XM_011523408.2
XP_011521710.1
3
291
XM_011523410.2
XP_011521712.1
3
209
XM_011523411.2
XP_011521713.1
4
447
XM_011523412.1
XP_011521714.1
4
417
XM_011523413.2
XP_011521715.1
3
849
XM_011523414.2
XP_011521716.1
3
635
XM_011523415.2
XP_011521717.1
2
166
XM_011523416.2
XP_011521718.1
3
374
XM_011523417.2
XP_011521719.1
3
291
XM_017023806.1
XP_016879295.1
2
180
XM_017023807.1
XP_016879296.1
3
291
XM_017023808.1
XP_016879297.1
4
446
XM_017023809.1
XP_016879298.1
2
166
XM_017023810.1
XP_016879299.1
3
634
GenBank mRNA
AB209829.1
-
2
142
AF153430.1
-
2
131
AK075036.1
-
3
215
AK131531.1
-
2
1794
AK290485.1
-
3
155
AK292351.1
-
3
213
AK295397.1
-
3
201
AK296631.1
-
2
139
AK298386.1
-
3
235
BC017342.2
-
3
177
4865641
BC025301.1
-
2
130
4637860
BC093028.1
-
3
214
CX867290.1
-
3
200
DB118903.1
-
2
162
DC362478.1
-
3
235
L33264.1
-
3
196
X78342.1
-
3
312
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
traversing start control point of mitotic cell cycle
negative regulation of cell proliferation
positive regulation of MAPK cascade
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0