Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075099.1 | NP_001068567.1 | 7 | 570 | ||
NM_003726.3 | NP_003717.3 | 7 | 570 | |||
XM_005257755.3 | XP_005257812.1 | 7 | 629 | |||
XM_017025257.1 | XP_016880746.1 | 6 | 492 | |||
XM_017025258.1 | XP_016880747.1 | 4 | 699 | |||
XM_017025259.1 | XP_016880748.1 | 8 | 707 | |||
XM_017025260.1 | XP_016880749.1 | 4 | 272 | |||
GenBank mRNA | AK290818.1 | - | 7 | 557 | ||
BC047870.1 | - | 7 | 542 | 5747375 | ||
DR007155.1 | - | 7 | 575 | |||
EU668346.1 | - | 7 | 542 | |||
Y11215.1 | - | 7 | 528 |