Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003737.3 | NP_003728.1 | 4 | 2475 | ||
GenBank mRNA | AB053446.1 | - | 4 | 2478 |