Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other B3GALNT1 SIRNA Assays ›
Gene Symbol:
B3GALNT1
Gene Name:
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases:
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3
Chromosome Location:
Chr.3: 161083883 - 161105528 on Build GRCh38
UniGene ID:
Hs.418062
Species:
Human
siRNA ID
HSS189510
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: B3GALNT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001038628.1
NP_001033717.1
8
1623
NM_003781.3
NP_003772.1
5
1300
NM_033167.2
NP_149357.1
6
1419
NM_033168.2
NP_149358.1
4
1205
NM_033169.2
NP_149359.1
5
1273
XM_005247859.4
XP_005247916.1
6
1363
XM_005247861.3
XP_005247918.1
7
1483
XM_005247862.3
XP_005247919.1
6
1341
XM_005247864.3
XP_005247921.1
5
1298
XM_006713803.3
XP_006713866.1
7
1482
XM_006713804.3
XP_006713867.1
4
1423
XM_006713805.3
XP_006713868.1
5
1542
XM_017007402.1
XP_016862891.1
6
1437
XM_017007403.1
XP_016862892.1
6
1385
XM_017007404.1
XP_016862893.1
6
1393
XM_017007405.1
XP_016862894.1
5
1266
XM_017007406.1
XP_016862895.1
7
1460
XM_017007407.1
XP_016862896.1
7
1609
XM_017007408.1
XP_016862897.1
6
1512
XM_017007409.1
XP_016862898.1
5
1405
XM_017007410.1
XP_016862899.1
7
1492
XM_017007411.1
XP_016862900.1
5
1414
XM_017007412.1
XP_016862901.1
4
1339
GenBank mRNA
AB050855.1
-
1
962
AB050856.1
-
5
1216
AB060691.1
-
1
631
AF132731.1
-
2
929
AF154848.1
-
5
1249
AK022491.1
-
5
1252
AK095917.1
-
4
1205
AK223439.1
-
5
1213
AY359049.1
-
4
1175
BC028571.1
-
8
1545
4838965
BC047618.1
-
5
1195
5259171
CR457170.1
-
1
854
Y15062.1
-
1
854
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
oligosaccharide biosynthetic process
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
galactosylgalactosylglucosylceramide beta-D-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline