Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251902.1 | NP_001238831.1 | 11 | 1949 | ||
NM_003985.4 | NP_003976.2 | 11 | 1934 | |||
XM_011524045.2 | XP_011522347.1 | 11 | 1982 | |||
XM_011524046.2 | XP_011522348.1 | 11 | 2065 | |||
XM_011524048.1 | XP_011522350.1 | 11 | 1839 | |||
GenBank mRNA | AK225403.1 | - | 11 | 1817 | ||
AK310684.1 | - | 12 | 1915 | |||
BC035782.1 | - | 10 | 1799 | 5589312 | ||
U43408.1 | - | 11 | 1833 |