Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251902.1 | NP_001238831.1 | 2 | 313 | ||
NM_003985.4 | NP_003976.2 | 2 | 313 | |||
XM_011524045.2 | XP_011522347.1 | 2 | 361 | |||
XM_011524046.2 | XP_011522348.1 | 2 | 450 | |||
XM_011524047.2 | XP_011522349.1 | 2 | 450 | |||
XM_011524048.1 | XP_011522350.1 | 2 | 196 | |||
GenBank mRNA | AK225403.1 | - | 2 | 196 | ||
AK310684.1 | - | 2 | 196 | |||
BC035782.1 | - | 1 | 178 | 5589312 | ||
DA089305.1 | - | 2 | 450 |