Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003810.3 | NP_003801.1 | 3 | 412 | ||
GenBank mRNA | AK312742.1 | - | 3 | 380 | ||
BC009795.1 | - | 3 | 309 | |||
BC032722.1 | - | 3 | 364 | 5504670 | ||
BT019564.1 | - | 3 | 289 | |||
CR456895.1 | - | 3 | 289 | |||
DA969653.1 | - | 3 | 412 | |||
U37518.1 | - | 3 | 376 | |||
U57059.1 | - | 3 | 379 |