Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003825.3 | NP_003816.2 | 7 | 948 | ||
NM_130798.2 | NP_570710.1 | 6 | 789 | |||
XM_006720725.2 | XP_006720788.1 | 8 | 1027 | |||
XM_017022692.1 | XP_016878181.1 | 8 | 1027 | |||
XM_017022693.1 | XP_016878182.1 | 7 | 654 | |||
XM_017022694.1 | XP_016878183.1 | 8 | 716 | |||
GenBank mRNA | AJ011915.1 | - | 7 | 570 | ||
AK290152.1 | - | 7 | 994 | |||
AK313161.1 | - | 7 | 592 | |||
AK315279.1 | - | 7 | 847 | |||
BC000148.2 | - | 7 | 580 | 2900640 | ||
BC003686.1 | - | 7 | 599 | 3461227 | ||
BC022890.1 | - | 7 | 604 | 4658930 | ||
BT006916.1 | - | 6 | 526 | |||
CR457212.1 | - | 6 | 526 | |||
DA650133.1 | - | 6 | 417 | |||
U55936.1 | - | 7 | 618 | |||
Y09567.1 | - | 6 | 526 | |||
Y09568.1 | - | 5 | 367 |