Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014238.1 | NP_055053.1 | 12 | 1522 | ||
XM_011525429.2 | XP_011523731.1 | 11 | 1674 | |||
XM_011525430.2 | XP_011523732.1 | 11 | 1676 | |||
XM_011525431.2 | XP_011523733.1 | 11 | 1676 | |||
XM_017025268.1 | XP_016880757.1 | 11 | 1674 | |||
XM_017025269.1 | XP_016880758.1 | 11 | 1674 | |||
XM_017025270.1 | XP_016880759.1 | 10 | 1409 | |||
XM_017025271.1 | XP_016880760.1 | 12 | 2381 | |||
XM_017025272.1 | XP_016880761.1 | 12 | 2364 | |||
XM_017025273.1 | XP_016880762.1 | 11 | 2187 | |||
XM_017025274.1 | XP_016880763.1 | 11 | 1390 | |||
XM_017025275.1 | XP_016880764.1 | 11 | 1377 | |||
XM_017025276.1 | XP_016880765.1 | 12 | 2329 | |||
XM_017025277.1 | XP_016880766.1 | 7 | 1072 | |||
XM_017025278.1 | XP_016880767.1 | 7 | 1082 | |||
GenBank mRNA | BC035488.1 | - | 13 | 2594 | ||
BC042106.1 | - | 12 | 1521 | |||
U43586.1 | - | 8 | 762 |