Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004665.4 | NP_004656.2 | 5 | 980 | ||
NM_078488.2 | NP_511043.1 | 6 | 808 | |||
NR_034173.2 | 4 | 691 | ||||
NR_110143.1 | 5 | 976 | ||||
NR_110144.1 | 5 | 768 | ||||
NR_110145.1 | 5 | 765 | ||||
XM_006715593.3 | XP_006715656.1 | 7 | 955 | |||
XM_011536231.1 | XP_011534533.1 | 5 | 876 | |||
XM_017011407.1 | XP_016866896.1 | 6 | 1591 | |||
XM_017011408.1 | XP_016866897.1 | 7 | 960 | |||
XM_017011409.1 | XP_016866898.1 | 7 | 853 | |||
GenBank mRNA | AB026705.1 | - | 5 | 862 | ||
AB435062.1 | - | 4 | 566 | |||
AJ132100.1 | - | 5 | 866 | |||
AK290908.1 | - | 5 | 876 | |||
AK292125.1 | - | 6 | 796 | |||
AK298719.1 | - | 5 | 661 | |||
BC126145.1 | - | 5 | 942 | 8991861 | ||
BC126147.1 | - | 5 | 942 | 8991863 | ||
D89974.1 | - | 5 | 866 |